Learn More
Cayman Chemical C14 Ceramide (d18:1/14:0), Purity: ≥95%, CAS: 34227-72-0, 25 mg
SDP
Catalog No. 502964131
Encompass_Preferred
A bioactive sphingolipid; levels increase time-dependently in a nutrient deprivation-induced necroptosis model in MEF cells; reduced levels in obese patients after chronic exercise correlate with weight and fat loss; higher levels in Parkinson’s disease patients correlate with memory deficits
Catalog No. 50-296-4131 Supplier Cayman Chemical Supplier No. 2253125 MG
A bioactive sphingolipid; levels increase time-dependently in a nutrient deprivation-induced necroptosis model in MEF cells; reduced levels in obese patients after chronic exercise correlate with weight and fat loss; higher levels in Parkinson’s disease patients correlate with memory deficits