Learn More
Medchemexpress LLC C14H11NO2S | 36341-25-0 | 99.3% | 257.31 | 50 MG
SDP
Catalog No. 5000387798
Encompass_Preferred
YL-109 is an antitumor agent that can induce carboxyl terminus of Hsp70-interacting protein (CHIP) expression through aryl hydrocarbon receptor (AhR) signaling. It has the ability to inhibit breast cancer cell growth and invasiveness.
  • Inhibits cell proliferation, motility, and invasiveness in breast cancer cells.
  • Increases both CHIP mRNA and protein levels in MDA-MB-231 cells.
  • Inhibits tumor growth and cancer metastasis of breast cancer cells in vivo.
Catalog No. 50-003-87798 Supplier Medchemexpress LLC Supplier No. HY1861950MG
YL-109 is an antitumor agent that can induce carboxyl terminus of Hsp70-interacting protein (CHIP) expression through aryl hydrocarbon receptor (AhR) signaling. It has the ability to inhibit breast cancer cell growth and invasiveness.
  • Inhibits cell proliferation, motility, and invasiveness in breast cancer cells.
  • Increases both CHIP mRNA and protein levels in MDA-MB-231 cells.
  • Inhibits tumor growth and cancer metastasis of breast cancer cells in vivo.