Learn More
Aviva Systems Biology Corp C14ORF101 Antibody - N-terminal region (ARP34357_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34357_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14ORF101. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C14ORF101. Alternate names: C14orf101, ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55352