Learn More
Aviva Systems Biology Corp C14orf104 Antibody - N-terminal region (ARP57125_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57125_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf104. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C14orf104. Alternate names: FLJ10563, KTU, CILD10, C14orf104ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62086