Learn More
Aviva Systems Biology Corp C14orf126 Antibody - middle region (ARP52511_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52511_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf126. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human C14orf126. Alternate names: MGC9912, C14orf126ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-723