Learn More
Aviva Systems Biology Corp C14orf129 Antibody - middle region (ARP56917_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56917_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf129. It was validated on Western Blot. C14orf129,chromosome 14 open reading frame 129,GSKIP; HSPC210; MGC4945; C14orf129,GSK3-beta interaction protein,GSK3-beta interaction protein,GSK3beta interaction proteinThe immunogen is a synthetic peptide directed towards the middle region of human C14orf129. Alternate names: GSKIP, HSPC210, MGC4945, C14orf129ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-236