Learn More
Aviva Systems Biology Corp C14orf130 Antibody - C-terminal region (ARP43243_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43243_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf130. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human C14orf130. Alternate names: FLJ10483, MGC9518, C14orf130ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58472