Learn More
Aviva Systems Biology Corp C14ORF131 Antibody - middle region (ARP32906_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32906_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C14ORF131. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C14ORF131. Alternate names: C14orf131ApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-014-54813