Learn More
Aviva Systems Biology Corp C14orf140 Antibody - N-terminal region (ARP53770_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53770_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf140. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C14orf140. Alternate names: FLJ23093, FAM164C, C14orf140ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-61122