Learn More
Aviva Systems Biology Corp C14orf148 Antibody - middle region (ARP52636_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52636_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf148. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C14orf148. Alternate names: FLJ32809, C14orf148ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse
Catalog No. 50-014-60790