Learn More
Aviva Systems Biology Corp RTRAF Antibody - middle region (ARP34546_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34546_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf166. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C14orf166. Alternate names: CGI-99, CGI99, CLE, CLE7, LCRP369, RLLM1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55443