Learn More
Aviva Systems Biology Corp RIOX1 Antibody - N-terminal region (ARP33580_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33580_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C14ORF169. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C14ORF169. Alternate names: NO66, MAPJDApplicationWB. Species reactivity: Cow, Dog, Human, Rabbit
Catalog No. 50-014-52910