Learn More
Aviva Systems Biology Corp C14orf172 Antibody - N-terminal region (ARP49042_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49042_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf172. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C14orf172. Alternate names: FLJ40452, GCD14, Gcd14p, TRM61, hTRM61, C14orf172ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60002