Learn More
Aviva Systems Biology Corp C14orf174 Antibody - N-terminal region (ARP51813_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51813_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf174. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C14orf174. Alternate names: FAM15A, FLJ35963, MGC132494, MGC132496, C14orf174ApplicationWB. Species reactivity: Horse, Human, Yeast
Catalog No. 50-014-53671