Learn More
Aviva Systems Biology Corp C14orf180 Antibody - N-terminal region (ARP44554_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44554_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf180. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C14orf180. Alternate names: FLJ38594, C14orf77ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58840