Learn More
Aviva Systems Biology Corp C14orf21 Antibody - middle region (ARP41178_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41178_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C14orf21. Alternate names: KIAA2021, C14orf21ApplicationWB. Species reactivity: Cow, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57751