Learn More
Aviva Systems Biology Corp C14orf28 Antibody - middle region (ARP51826_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51826_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf28. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human C14orf28. Alternate names: DRIP1, c14_5270ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-599