Learn More
Aviva Systems Biology Corp ARMH4 Antibody - N-terminal region (ARP44413_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44413_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf37. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C14orf37. Alternate names: c14_5376ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-014-58802