Learn More
Aviva Systems Biology Corp CCDC197 Antibody - N-terminal region (ARP55545_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55545_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf48. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human C14orf48. Alternate names: DKFZp434O1910, MGC33356, c14_5713, C14orf48ApplicationWB. Species reactivity: Human
Catalog No. 50-286-054