Learn More
Cayman Chemical C15 Ceramide (d18:1/15:0), Purity: ≥98%, CAS: 67492-15-3, 10 mg
SDP
Catalog No. 502936198
Encompass_Preferred
A naturally occurring ceramide; has been found in L. nidifica and A. glomerata extracts, human erythrocytes, and the stratum corneum of P. domesticus
Catalog No. 50-293-6198 Supplier Cayman Chemical Supplier No. 2439310 MG
A naturally occurring ceramide; has been found in L. nidifica and A. glomerata extracts, human erythrocytes, and the stratum corneum of P. domesticus