Learn More
Aviva Systems Biology Corp CFAP161 Antibody - C-terminal region (ARP53271_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53271_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C15orf26. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C15orf26. Alternate names: FLJ38615ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig
Catalog No. 50-014-60986