Learn More
Aviva Systems Biology Corp TMEM266 Antibody - middle region (ARP52897_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52897_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C15orf27. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C15orf27. Alternate names: FLJ30304, FLJ38190ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60862