Learn More
Aviva Systems Biology Corp C15orf48 Antibody - N-terminal region (ARP50069_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50069_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C15orf48. It was validated on Western Blot. . Alternate names: FLJ22645, FOAP-11, MGC32925, NMES1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-480