Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C16 ceramide-1-phosphate (d18:1/16:0) ammonium | 1246303-22-9 | 98.0% | 634.91 | C34H71N2O6P | 5 MG
SDP

Catalog No. 5000221646
Encompass_Preferred

C16 ceramide-1-phosphate (d18:1/16:0) (ammonium) is the ammonium salt of a bioactive sphingolipid, supplied as a purified research reagent for biochemical and cell biology studies of sphingolipid signaling and metabolism. It is provided as a characterized dry material for laboratory research use only and requires protected cold storage.

  • High purity (98.0%).
  • Molecular weight 634.91.
  • Chemical formula C34H71N2O6P.
  • Available in small pack sizes such as 5 MG.
  • Storage: in solvent, -80°C (6 months) or -20°C (1 month); protect from light and store under nitrogen.
  • Intended for biochemical assays and cell biology research; not for human use.

Catalog No. 50-002-21646 Supplier Medchemexpress LLC Supplier No. HY1454675MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C16 ceramide-1-phosphate (d18:1/16:0) (ammonium) is the ammonium salt of a bioactive sphingolipid, supplied as a purified research reagent for biochemical and cell biology studies of sphingolipid signaling and metabolism. It is provided as a characterized dry material for laboratory research use only and requires protected cold storage.

  • High purity (98.0%).
  • Molecular weight 634.91.
  • Chemical formula C34H71N2O6P.
  • Available in small pack sizes such as 5 MG.
  • Storage: in solvent, -80°C (6 months) or -20°C (1 month); protect from light and store under nitrogen.
  • Intended for biochemical assays and cell biology research; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.