Learn More
Medchemexpress LLC C16-ceramide | 24696-26-2 | MFCD00056205 | ≥99.0% | 537.90 | C34H67NO3 | 5 MG
SDP
Catalog No. 5000212698
Encompass_Preferred
C16-ceramide is a natural ceramide lipid reagent used in cell biology research. It binds directly and selectively to p53 to activate p53 signaling, can induce apoptosis in certain cell lines, and is used to probe ceramide-mediated regulatory pathways. Provided as solid material and as solution formulations for assay compatibility.
  • Direct activation of p53 through selective binding.
  • Induces apoptosis in p53-related cellular assays.
  • High purity (≥99.0%) suitable for research use.
  • Available as solid and solution formats for flexibility.
  • Stable when stored protected from light at recommended temperatures.
Catalog No. 50-002-12698 Supplier Medchemexpress LLC Supplier No. HY1003545MG
C16-ceramide is a natural ceramide lipid reagent used in cell biology research. It binds directly and selectively to p53 to activate p53 signaling, can induce apoptosis in certain cell lines, and is used to probe ceramide-mediated regulatory pathways. Provided as solid material and as solution formulations for assay compatibility.
  • Direct activation of p53 through selective binding.
  • Induces apoptosis in p53-related cellular assays.
  • High purity (≥99.0%) suitable for research use.
  • Available as solid and solution formats for flexibility.
  • Stable when stored protected from light at recommended temperatures.