Learn More
Medchemexpress LLC C16-ceramide | 24696-26-2 | MFCD00056205 | >=99.0% | 537.90 g/mol | C34H67NO3 | 50 MG
SDP
Catalog No. 5000213278
Encompass_Preferred
C16-ceramide is a naturally occurring ceramide that activates the tumor suppressor p53 via direct, selective binding to its core domain. Supplied as a high-purity solid, it is used as a biochemical research reagent to study p53 signaling, apoptosis, and sphingolipid metabolism in cell-based and biochemical assays.
  • Activates p53 through direct binding.
  • Induces apoptosis and decreases cell viability in cell-based assays.
  • High purity suitable for research applications.
  • Solid, white to off-white appearance with defined storage recommendations.
  • Available in small pack sizes for laboratory use.
Catalog No. 50-002-13278 Supplier Medchemexpress LLC Supplier No. HY10035450MG
C16-ceramide is a naturally occurring ceramide that activates the tumor suppressor p53 via direct, selective binding to its core domain. Supplied as a high-purity solid, it is used as a biochemical research reagent to study p53 signaling, apoptosis, and sphingolipid metabolism in cell-based and biochemical assays.
  • Activates p53 through direct binding.
  • Induces apoptosis and decreases cell viability in cell-based assays.
  • High purity suitable for research applications.
  • Solid, white to off-white appearance with defined storage recommendations.
  • Available in small pack sizes for laboratory use.