Learn More
Enzo Life Sciences C16 Ceramide (25 mg). CAS: 24696-26-2
SDP
Supplier:  Enzo Life Sciences BMLSL1150025
Encompass_Preferred
An abundant molecular species of endogenous ceramide. It induces phosphorylation of epidermal growth factor receptor on Thr669 in A-431 cells by stimulation of ceramide-activated protein kinase and activates NF-κB at concentrations as low as 50 nM. Activates PKCζ. Alternative name: N-Palmitoyl-D-erythro-sphingosine, N-Hexadecanoyl-D-erythro-sphingosine, N-Palmitoyl ceramide. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol (5mg/ml warm). Long Term Storage: -20°C.
Catalog No. 50-201-2112