Learn More
Cayman Chemical C16 Ceramide (d18:1/16:0), Purity: ≥98%, CAS: 24696-26-2, 10 mg
SDP
Catalog No. 502891254
Encompass_Preferred
A sphingolipid; levels are increased by PPMP or MAPP in isolated human neutrophils and are correlated with an increase in apoptosis; accumulates in the lungs of patients with cystic fibrosis compared with the lungs of patients with pulmonary hypertension; plasma levels are increased in patients with acute coronary syndrome, and an increased plasma ratio of C16 to C24:0 ceramide is associated with major adverse cardiac events; levels are increased in tumor tissue from patients with breast cancer and is associated with metastatic cancer
Catalog No. 50-289-1254 Supplier Cayman Chemical Supplier No. 1068110 MG
A sphingolipid; levels are increased by PPMP or MAPP in isolated human neutrophils and are correlated with an increase in apoptosis; accumulates in the lungs of patients with cystic fibrosis compared with the lungs of patients with pulmonary hypertension; plasma levels are increased in patients with acute coronary syndrome, and an increased plasma ratio of C16 to C24:0 ceramide is associated with major adverse cardiac events; levels are increased in tumor tissue from patients with breast cancer and is associated with metastatic cancer