Learn More
Medchemexpress LLC C16-ceramide-d31 | 852043-41-5 | MFCD22416906 | 98.0% | 569.09 | C34H36D31NO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY100354S41MG
Encompass_Preferred
C16-ceramide-d31 is a deuterium-labeled analogue of C16-ceramide intended for use as an internal standard or tracer in lipidomics and analytical research. The isotopically labeled compound enables reliable quantitation and metabolic tracing in mass spectrometry-based assays.
  • Isotopically labeled with deuterium (d31).
  • Suitable as an internal standard or tracer for lipidomics and mass spectrometry.
  • High purity: 98.0%.
  • Available in small research quantities (e.g., 1 MG).
  • Molecular formula C34H36D31NO3 and molecular weight 569.09.
Catalog No. 50-002-14353