Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C16 Ceramide-d7 (d16:1,C16:0) | 1840942-13-3 | 99.9% | 544.94 | 1 MG
SDP

Catalog No. 5000437814
Encompass_Preferred

C16 Ceramide-d7 (d16:1,C16:0) is the deuterium labeled C16 Ceramide (d16:1,C16:0).

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.

Catalog No. 50-004-37814 Supplier Medchemexpress LLC Supplier No. HY116258S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C16 Ceramide-d7 (d16:1,C16:0) is the deuterium labeled C16 Ceramide (d16:1,C16:0).

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.