Learn More
Avanti Polar Lipids N-palmitoyl-D-erythro-sphingosine (d7), 1840942-13-3, 1 mg
Catalog No. NC1290251
Encompass
Molecular Formula: C34H60D7NO3, Formula Weight: 544.944, Purity >99%. Monitoring the levels of the ceramides (Cer) d18:1/16:0, Cer d18:1/18:0, Cer d18:1/24:0, and Cer d18:1/24:1 and ratios thereof in human plasma empowers the prediction of fatal outcome of coronary artery disease.
Catalog No. NC1290251 Supplier Avanti Polar Lipids Supplier No. 860676P1MG
Molecular Formula: C34H60D7NO3, Formula Weight: 544.944, Purity >99%. Monitoring the levels of the ceramides (Cer) d18:1/16:0, Cer d18:1/18:0, Cer d18:1/24:0, and Cer d18:1/24:1 and ratios thereof in human plasma empowers the prediction of fatal outcome of coronary artery disease.