Learn More
Cayman Chemical C16 CeramIde-d7d18 1-d7161mg
SDP
Catalog No. 502995478
Encompass_Preferred
An internal standard for the quantification of C16 ceramide by GC- or LC-MS
Catalog No. 50-299-5478 Supplier Cayman Chemical Supplier No. 227871 MG
An internal standard for the quantification of C16 ceramide by GC- or LC-MS