Learn More
Cayman Chemical C16 CeramIde-d7d18 1-d716500ug
SDP
Catalog No. 502995456
Encompass_Preferred
An internal standard for the quantification of C16 ceramide by GC- or LC-MS
Catalog No. 50-299-5456 Supplier Cayman Chemical Supplier No. 22787500 UG
An internal standard for the quantification of C16 ceramide by GC- or LC-MS