Learn More
Medchemexpress LLC C16 ceramide (N-palmitoylsphingosine, D-erythro) | 24696-26-2 | MFCD00056205 | 98.0% | 537.90 | C34H67NO3 | 25 MG
SDP
Catalog No. 5000213277
Encompass_Preferred
C16 ceramide is a naturally occurring sphingolipid (N-palmitoylsphingosine) used as a research reagent that activates p53 through direct, selective binding. Supplied as a white to off-white solid, it is used in cell biology studies of apoptosis, cell signaling, and mitochondrial membrane permeability. Store protected from light at 4°C; in solution store at -80°C (long term) or -20°C (short term).
  • Activates p53 through direct, selective binding.
  • Endogenous sphingolipid for studies of apoptosis and cell signaling.
  • Supplied as a stable solid, white to off-white in color.
  • Compatible with common organic solvents; prepare fresh solutions and protect from light.
  • Intended for research use only.
Catalog No. 50-002-13277 Supplier Medchemexpress LLC Supplier No. HY10035425MG
C16 ceramide is a naturally occurring sphingolipid (N-palmitoylsphingosine) used as a research reagent that activates p53 through direct, selective binding. Supplied as a white to off-white solid, it is used in cell biology studies of apoptosis, cell signaling, and mitochondrial membrane permeability. Store protected from light at 4°C; in solution store at -80°C (long term) or -20°C (short term).
  • Activates p53 through direct, selective binding.
  • Endogenous sphingolipid for studies of apoptosis and cell signaling.
  • Supplied as a stable solid, white to off-white in color.
  • Compatible with common organic solvents; prepare fresh solutions and protect from light.
  • Intended for research use only.