Learn More
Cayman Chemical C16 dihydro Ceramide-d9 (d18:0/16:0-d9), Purity: ≥99% deuterated forms (d1-d9), CAS: 2260669-52-9, 1 mg
SDP
Catalog No. 502915788
Encompass_Preferred
An internal standard for the quantification of C16 dihydro ceramide by GC- or LC-MS
Catalog No. 50-291-5788 Supplier Cayman Chemical Supplier No. 393891 MG
An internal standard for the quantification of C16 dihydro ceramide by GC- or LC-MS