Learn More
Cayman Chemical C16 dIhydro CeramIded18 01 1mg
SDP
Catalog No. 503016030
Encompass_Preferred
A precursor in the de novo synthesis of C16 ceramide; inhibits C16 ceramide-induced membrane permeabilization in rat liver mitochondria and channel formation in liposomes; biologically inactive as a single agent
Catalog No. 50-301-6030 Supplier Cayman Chemical Supplier No. 243691 MG
A precursor in the de novo synthesis of C16 ceramide; inhibits C16 ceramide-induced membrane permeabilization in rat liver mitochondria and channel formation in liposomes; biologically inactive as a single agent