Learn More
Cayman Chemical C16 dIhydro CeramIded9d18500ug
SDP
Supplier:  Cayman Chemical 39389500 UG
Encompass_Preferred
An internal standard for the quantification of C16 dihydro ceramide by GC- or LC-MS
Catalog No. 50-291-7052