Learn More
Cayman Chemical C16 dIhydro CRamIDd18 01 10mg
SDP
Catalog No. 503015785
Encompass_Preferred
A precursor in the de novo synthesis of C16 ceramide; inhibits C16 ceramide-induced membrane permeabilization in rat liver mitochondria and channel formation in liposomes; biologically inactive as a single agent
Catalog No. 50-301-5785 Supplier Cayman Chemical Supplier No. 2436910 MG
A precursor in the de novo synthesis of C16 ceramide; inhibits C16 ceramide-induced membrane permeabilization in rat liver mitochondria and channel formation in liposomes; biologically inactive as a single agent