Learn More
Cayman Chemical C16 Globotriaosylceramide d1
SDP
Catalog No. 503015867
Encompass_Preferred
A sphingolipid; acts as a receptor for Shiga toxin in B cell-derived Raji cells and THP-1 monocytes; accumulates in various cells in patients with Fabry disease; upregulated in plasma of patients with ovarian carcinoma compared to those with benign ovarian tumors or uterine fibroids
Catalog No. 50-301-5867 Supplier Cayman Chemical Supplier No. 24875500 UG
A sphingolipid; acts as a receptor for Shiga toxin in B cell-derived Raji cells and THP-1 monocytes; accumulates in various cells in patients with Fabry disease; upregulated in plasma of patients with ovarian carcinoma compared to those with benign ovarian tumors or uterine fibroids