Learn More
Cayman Chemical C16 Glucosylceramide-d3 (d18:1/16:0-d3), Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502934871
Encompass_Preferred
An internal standard for the quantification of C16 glucosylceramide by GC- or LC-MS
Catalog No. 50-293-4871 Supplier Cayman Chemical Supplier No. 246211 MG
An internal standard for the quantification of C16 glucosylceramide by GC- or LC-MS