Learn More
AdipoGen C16-L-HSL
SDP
Supplier:  AdipoGen CDXH0113M050
Encompass_Preferred
Chemical. CAS 87206-01-7. Formula C20H37NO3. MW 339.5. Synthetic. N-Hexadecanoyl-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. C16-HSL is a lipophilic, long acyl side-chain bearing AHL, produced by the LuxI AHL synthase homolog SinI involved in quorum-sensing signaling in S. meliloti, a nitrogen-fixing bacterial symbiont of certain legumes. C16-HSL is the most abundant AHL produced by the proteobacterium R. capsulatus and activates genetic exchange between R. capsulatus cells. It tends to localize in relatively lipophilic cellular environments of bacteria and cannot diffuse freely through the cell membrane. The long-chain N-acylhomoserine lactones may be exported from cells by efflux pumps or may be transported between communicating cells by extracellular outer membrane vesicles.
Catalog No. 50-256-0723