Learn More
Cayman Chemical C16 Lactosylceramide-d3 (d18:1/16:0-d3), Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502936404
Encompass_Preferred
An internal standard for the quantification of C16 lactosylceramide by GC- or LC-MS
Catalog No. 50-293-6404 Supplier Cayman Chemical Supplier No. 246251 MG
An internal standard for the quantification of C16 lactosylceramide by GC- or LC-MS