Learn More
Cayman Chemical C16 Ceramide-mPEG(2000), Purity: ≥90%, CAS: 212116-78-4, 1 mg
SDP
Catalog No. 502943333
Encompass_Preferred
A PEGylated form of C16 ceramide; has been used in the generation of microspheres, LNPs, and liposomes for the delivery of small molecules, mRNA, and plasmid DNA in vitro and in vivo; liposomes containing C16 ceramide-mPEG(2000) with a stearylated-INF7 and have been used to transfect plasmid DNA into MFLM-4 cells; microspheres containing C16 ceramide-mPEG(2000) and encapsulating oxaliplatin increase survival and decrease the number of small intestine, colon, and cecum polyps in an Apc+/Minz spontaneous colorectal adenoma mouse model
Catalog No. 50-294-3333 Supplier Cayman Chemical Supplier No. 419551 MG
A PEGylated form of C16 ceramide; has been used in the generation of microspheres, LNPs, and liposomes for the delivery of small molecules, mRNA, and plasmid DNA in vitro and in vivo; liposomes containing C16 ceramide-mPEG(2000) with a stearylated-INF7 and have been used to transfect plasmid DNA into MFLM-4 cells; microspheres containing C16 ceramide-mPEG(2000) and encapsulating oxaliplatin increase survival and decrease the number of small intestine, colon, and cecum polyps in an Apc+/Minz spontaneous colorectal adenoma mouse model