Learn More
Medchemexpress LLC C16 PEG2000 Ceramide | 212116-78-4 | 99.37% | (C2H4O)nC39H73NO6 | 25 MG
SDP
Catalog No. 5000356531
Encompass_Preferred
C16 PEG2000 Ceramide is a polyethylene glycolylated ceramide that functions as a lipid carrier for delivery and induces autophagy. This compound is also utilized in cancer research, exhibiting cytotoxicity and promoting autophagy in N2a cells in vitro.
  • Functions as a lipid carrier for delivery
  • Induces autophagy
  • Used in cancer research
  • Exhibits cytotoxicity in N2a cells
  • Promotes autophagy in N2a cells
  • Increases LC3-II/LC3-I ratios in N2a cells
  • Reduces WT-Tau and P301L-Tau proteins in N2a cells
Catalog No. 50-003-56531 Supplier Medchemexpress LLC Supplier No. 25MGHY144005
C16 PEG2000 Ceramide is a polyethylene glycolylated ceramide that functions as a lipid carrier for delivery and induces autophagy. This compound is also utilized in cancer research, exhibiting cytotoxicity and promoting autophagy in N2a cells in vitro.
  • Functions as a lipid carrier for delivery
  • Induces autophagy
  • Used in cancer research
  • Exhibits cytotoxicity in N2a cells
  • Promotes autophagy in N2a cells
  • Increases LC3-II/LC3-I ratios in N2a cells
  • Reduces WT-Tau and P301L-Tau proteins in N2a cells