Learn More
Medchemexpress LLC C16 PEG2000 ceramide | 212116-78-4 | MFCD02265697 | 99.4% | (C2H4O)nC39H73NO6 | 10MG
SDP
Catalog No. 5000259153
Encompass_Preferred
C16 PEG2000 ceramide is a polyethylene glycol-conjugated C16 ceramide (PEG MW ≈2000) used as a PEGylated lipid in nanoparticle and liposome formulations to improve stability and delivery of hydrophobic agents. It is typically supplied as a white to off-white powder with reported high purity.
  • Pegylated C16 ceramide for lipid formulations.
  • Enhances stability of lipid nanoparticles and liposomes.
  • Improves solubility and dispersibility of hydrophobic compounds.
  • White to off-white powder form for ease of handling.
  • High purity suitable for research applications.
Catalog No. 50-002-59153 Supplier Medchemexpress LLC Supplier No. HY14400510MG
C16 PEG2000 ceramide is a polyethylene glycol-conjugated C16 ceramide (PEG MW ≈2000) used as a PEGylated lipid in nanoparticle and liposome formulations to improve stability and delivery of hydrophobic agents. It is typically supplied as a white to off-white powder with reported high purity.
  • Pegylated C16 ceramide for lipid formulations.
  • Enhances stability of lipid nanoparticles and liposomes.
  • Improves solubility and dispersibility of hydrophobic compounds.
  • White to off-white powder form for ease of handling.
  • High purity suitable for research applications.