Learn More
Cayman Chemical C16 Phytoceramide (t18:0/16:0), Purity: ≥98%, CAS: 111149-09-8, 5 mg
SDP
Catalog No. 502993930
Encompass_Preferred
A phytoceramide; levels increase following heat stress in S. cerevisiae; used in stratum cornea substitutes to study percutaneous penetration in vitro
Catalog No. 50-299-3930 Supplier Cayman Chemical Supplier No. 228265 MG
A phytoceramide; levels increase following heat stress in S. cerevisiae; used in stratum cornea substitutes to study percutaneous penetration in vitro