Learn More
Abcam C16, PKR inhibitor, 25MG
Catalog No. 502705591
Encompass_Preferred
MW 268.3 Da, Purity >98%. Specific double-stranded RNA-dependent protein kinase (PKR) inhibitor. Prevents PKR-induced neuronal loss in vivo. Inhibits apoptotic PKR/eIF2α signaling pathway in vivo. Neuroprotective, antitumor and anti-inflammatory agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5591 Supplier Abcam Supplier No. AB14459525MG
Add to Cart
Add to Cart
MW 268.3 Da, Purity >98%. Specific double-stranded RNA-dependent protein kinase (PKR) inhibitor. Prevents PKR-induced neuronal loss in vivo. Inhibits apoptotic PKR/eIF2α signaling pathway in vivo. Neuroprotective, antitumor and anti-inflammatory agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement