Learn More
Cayman Chemical C16 Sphingomyelin (d18:1/16:0), Purity: ≥98%, CAS: 6254-89-3, 5 mg
SDP
Catalog No. 502890080
Encompass_Preferred
A naturally occurring sphingolipid; levels are elevated in the prefrontal cortex and cerebellum in a rat model of STZ-induced diabetes, as well as in the liver in mouse models of Niemann-Pick type C1 disease; plasma are decreased in patients with prostate cancer; has been used in model membranes with POPC and cholesterol in the study of lipid raft dynamics and membrane organization
Catalog No. 50-289-0080 Supplier Cayman Chemical Supplier No. 100079465 MG
A naturally occurring sphingolipid; levels are elevated in the prefrontal cortex and cerebellum in a rat model of STZ-induced diabetes, as well as in the liver in mouse models of Niemann-Pick type C1 disease; plasma are decreased in patients with prostate cancer; has been used in model membranes with POPC and cholesterol in the study of lipid raft dynamics and membrane organization