Learn More
Alkali Scientific Alkali Scientific™ C1612 MC-12, 12-Place High-Speed Bench Top Micro-Centrifuge
SDP
Catalog No. 503029609
Encompass_Preferred
The Alkali Scientific™ B3D1008 Mini Blot Boy™ 3D orbital shaker is a simple and affordable shaker for all your blot related applications. The tilt angle and speed have been optimally set for gentle, but thorough mixing in gel trays, boxes and other flat vessels.
Catalog No. 50-302-9609 Supplier Alkali Scientific Supplier No. RC1612
The Alkali Scientific™ B3D1008 Mini Blot Boy™ 3D orbital shaker is a simple and affordable shaker for all your blot related applications. The tilt angle and speed have been optimally set for gentle, but thorough mixing in gel trays, boxes and other flat vessels.