Learn More
Cayman Chemical C16 (2'(S)-hydroxy) Ceramide (d18:1/16:0), Purity: ≥95%, CAS: 890041-50-6, 1 mg
SDP
Catalog No. 502986817
Encompass_Preferred
A synthetic ceramide; has no effect on C6 glioma cell viability at concentrations up to 20 µM
Catalog No. 50-298-6817 Supplier Cayman Chemical Supplier No. 225161 MG
A synthetic ceramide; has no effect on C6 glioma cell viability at concentrations up to 20 µM